
Fensulfothion oxon

Synonym(s):
Diethyl 4-(methylsulfinyl)phenyl phosphate, Fensulfothion PO-sulfoxide, NSC 133045
Empirical Formula (Hill Notation):
C11H17O5PS
CAS Number:
Molecular Weight:
292.29
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing