
Famciclovir Related Compound A

Synonym(s):
2-[2-(2-Amino-9H-purin-9-yl)ethyl]propane-1,3-diol monohydrochloride
Empirical Formula (Hill Notation):
C10H15N5O2 · HCl
CAS Number:
Molecular Weight:
273.72
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2