
(−)-Epigallocatechin

Synonym(s):
(−)-cis-2-(3,4,5-Trihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol, (−)-cis-3,3′,4′,5,5′,7-Hexahydroxyflavane, (−)-Epigallocatechin
Empirical Formula (Hill Notation):
C15H14O7
CAS Number:
Molecular Weight:
306.27
Products
Product Category
Brand
Biological Source
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4