
D-Saccharic acid 1,4-lactone monohydrate

Synonym(s):
D-Glucaric acid-1,4-lactone, D-Saccharolactone
Empirical Formula (Hill Notation):
C6H8O7 · H2O
CAS Number:
Molecular Weight:
210.14
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing