
Donepezil related compound A

Synonym(s):
(E)-2-[(1-Benzylpiperidin-4-yl)methylene]-5,6-dimethoxyindan-1-one
Empirical Formula (Hill Notation):
C24H27NO3
CAS Number:
Molecular Weight:
377.48
Products
Formula Weight
Manufacturer
Application
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing