
Delphinidin 3-sambubioside chloride

Synonym(s):
Delphinidin 3-O-(2-O-β-D-xylopyranosyl)-β-D-glucopyranoside chloride
Empirical Formula (Hill Notation):
C26H29O16Cl
CAS Number:
Molecular Weight:
632.95
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing