Skip to Content
Merck
D-(−)-Arabinose suitable for microbiology, ≥99.0%

D-(−)-Arabinose

D-(−)-Arabinose suitable for microbiology, ≥99.0%
Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
EC Number:

Products

Product Category
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing