
Dactinomycin

Synonym(s):
Actinomycin D, Actinomycin C1, Actinomycin IV, Dactinomycin
Empirical Formula (Hill Notation):
C62H86N12O16
CAS Number:
Molecular Weight:
1255.42
Products
Product Category
Color
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing