
Chloroquine Related Compound A

Synonym(s):
4,7-Dichloroquinoline
Empirical Formula (Hill Notation):
C9H5Cl2N
CAS Number:
Molecular Weight:
198.05
Products
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2