
11β,16α,17,21-Tetrahydroxypregna-1,4-dien-3,20-dione

Synonym(s):
16α-Hydroxyprednisolone
Empirical Formula (Hill Notation):
C21H28O6
CAS Number:
Molecular Weight:
376.44
Products
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade
Showing 1-1 of 1