Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.15106

Sigma-Aldrich

2-Amino-4,6-dichlorobenzoic acid

for synthesis

Synonym(s):

2-Amino-4,6-dichlorobenzoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$605.00

$605.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$605.00

About This Item

Empirical Formula (Hill Notation):
C7H5Cl2NO2
CAS Number:
Molecular Weight:
206.03
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

$605.00


Estimated to ship onMay 18, 2025


Request a Bulk Order

Quality Level

form

powder

pH

2.3 (20 °C in H2O)

solubility

1.25 g/L

storage temp.

2-30°C

SMILES string

Clc1c(c(cc(c1)Cl)N)C(=O)O

InChI

1S/C7H5Cl2NO2/c8-3-1-4(9)6(7(11)12)5(10)2-3/h1-2H,10H2,(H,11,12)

InChI key

VNOYJBSEORILIE-UHFFFAOYSA-N

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
8.188348.21989399760
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

form

powder

form

powder

form

liquid

form

-

pH

2.3 (20 °C in H2O)

pH

-

pH

-

pH

-

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service