Merck
  • Influence of drug transporter polymorphisms on pravastatin pharmacokinetics in humans.
Influence of drug transporter polymorphisms on pravastatin pharmacokinetics in humans.
Pharmaceutical research (2006-12-21)
Kari T Kivistö, Mikko Niemi
ABSTRACT
The role of drug transporters in pravastatin disposition is underlined by the fact that pravastatin does not undergo significant cytochrome P-450 (CYP)-mediated biotransformation. The organic anion transporting polypeptide 1B1 (OATP1B1), encoded by SLCO1B1, and multidrug resistance-associated protein 2 [MRP2 (ABCC2)], are thought to be the major transporters involved in the pharmacokinetics of pravastatin in humans. Other transporters that may play a role include OATP2B1, organic anion transporter 3 (OAT3), bile salt export pump (BSEP), and the breast cancer resistance protein (BCRP). OATP1B1 and MRP2 mediate the hepatic uptake and biliary excretion of pravastatin, respectively. The SLCO1B1 and ABCC2 polymorphisms probably contribute to the high interindividual variability in pravastatin disposition. Recent small studies have characterized the impact of the SLCO1B1 polymorphism on pravastatin in humans, and especially the c.521T>C single-nucleotide polymorphism (SNP) seems to be an important determinant of pravastatin pharmacokinetics. Pravastatin plasma concentrations may be up to 100% higher in subjects carrying the c.521C variant, as found in the *5, *15, *16, and *17 haplotypes, reflecting diminished OATP1B1-mediated uptake into the major site of pravastatin elimination, the liver. The SLCO1B1 polymorphism seems to have a similar impact on the pharmacokinetics of single- and multiple-dose pravastatin. Overall, 2-5% of individuals in various populations may be expected to show markedly elevated plasma pravastatin concentrations due to the SLCO1B1 polymorphism. Of note, the impact of the SLCO1B1 polymorphism on statins may be dependent on ethnicity. Although individuals with a diminished hepatic uptake of pravastatin might be expected to show reduced cholesterol-lowering efficacy due to lower intracellular pravastatin concentrations, there is preliminary evidence to suggest that the SLCO1B1 polymorphism is not a major determinant of non-response to pravastatin. The possible consequences of drug transporter polymorphisms, especially the SLCO1B1 and ABCC2 polymorphisms, for the lipid-lowering efficacy and tolerability of pravastatin in various ethnic groups warrant further study.
Sigma-Aldrich
SB-BSEP-HEK293
Sigma-Aldrich
SB-mouseBsep-Sf9, membrane preparation for Vesicular Transport Assays, recombinant, expressed in baculovirus infected Sf9 cells
Sigma-Aldrich
SB-BSEP-Hi5, Membrane preparation for Vesicular Transport Assays, recombinant, expressed in Baculovirus infected High-5 cells