Merck
Electrospray ionization tandem mass spectrometry of chaetoglobosins.
Rapid communications in mass spectrometry : RCM (2012-08-14)
Guo-Bo Xu, Li-Mei Li, Dong-Mei Fang, Guo-You Li, Guo-Lin Zhang, Zhi-Jun Wu
ABSTRACT
Chaetoglobosins are a family of macrocyclic polyketide alkaloids. They possess many similar isomers and exhibit a wide range of biological activities. Thus, there is a need for reliable, fast, and low-cost analysis of this class of compounds. A series of seven chaetoglobosins from Chaetomium globosum, including two types of isomers, were investigated using electrospray ionization quadrupole time-of-flight tandem mass spectrometry (ESI-QTOF-MS/MS) in both positive- and negative-ion mode. The identity of major product ions was supported by deuterium-labeling experiments. In positive-ion mode, the product ion at m/z 130 is the characteristic ion of the indolyl group. A McLafferty rearrangement might play a significant role in the fragmentation of the macrocycle moiety for most chaetoglobosins and produces two series of characteristic product ions, accompanied by neutral losses. The characteristic product ion at m/z 309 in the MS/MS spectrum of chaetoglobosins E indicates the structure of the cyclic olefinic bond in ring B and can be used to distinguish it from the isomers, chaetoglobosins F(ex) , which has an exocyclic double bond on ring B. In negative-ion mode, the McLafferty rearrangement has an important role in the fragmentation pattern of the macrocycle. Some high-abundance radical ions were detected. The radical product ion at m/z 138 might differentiate chaetoglobosins F and penochalasin F, isomers which have very similar structures. In summary, complementary information obtained from fragmentation experiments of [M+H](+) and [M-H](-) precursor ions is especially valuable for rapid identification of chaetoglobosins. The high-abundance radical ions in negative-ion mode are also of scientific interest.
Sigma-Aldrich
Chaetoglobosin A, from Chaetomium globosum, ≥98% (HPLC)