Merck
  • Narrowband ultraviolet photodetector based on MgZnO and NPB heterojunction.
Narrowband ultraviolet photodetector based on MgZnO and NPB heterojunction.
Optics letters (2012-08-04)
Zuofu Hu, Zhenjun Li, Lu Zhu, Fengjuan Liu, Yanwu Lv, Xiqing Zhang, Yongsheng Wang
ABSTRACT
An ultraviolet photodetector was fabricated based on Mg0.07Zn0.93O heterojunction. N, N'-bis (naphthalen-1-y1)-N, N'-bis(pheny) benzidine was selected as the hole transporting layer. I-V characteristic curves of the device were measured in the dark and under the illumination of 340 nm UV light with density of 1.33 mW/cm2. The device showed a low dark current of about 3×10(-10) A and a high photo-dark current ratio of 1×10(5) at -2 V bias. A narrowband photoresponse was observed from 300 to 400 nm and centered at 340 nm with a full width at half-maximum of only 30 nm. The maximum peak response is at 340 nm, which is 0.192 A/W at the bias of -1 V.
Product Description
Supelco
1-Naphthylamine, analytical standard
Sigma-Aldrich
1-Naphthylamine, 97%
Millipore
Nitrate Reagent A, suitable for microbiology