Merck
An organocatalytic asymmetric chlorolactonization.
Journal of the American Chemical Society (2010-02-23)
Daniel C Whitehead, Roozbeh Yousefi, Arvind Jaganathan, Babak Borhan
ABSTRACT
A reagent controlled organocatalytic enantioselective chlorolactonization reaction has been developed. Several 4-aryl pentenoic acids were cyclized in the presence of 0.1 equiv of (DHQD)(2)PHAL, employing various N-chlorinated hydantoins as the terminal chlorenium source. Ten examples are presented with selectivities ranging from 43 to 90% ee. This work represents the first example of an enantioselective reagent-controlled chlorolactonization that approaches synthetically useful enantioselectivities.
Sigma-Aldrich
4-Pentenoic acid, ≥98%, stabilized, FG
Sigma-Aldrich
4-Pentenoic acid, 97%