Merck
Chemical constituents from aerial part of Curcuma wenyujin.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2008-03-15)
Zheng-Ming Tao, Ying-Yu Li, Ping Ji, Yu-Bo Wang, Guo-Wei Qin
ABSTRACT
To investigate the chemical constituents from aerial part of Curcuma wenyujin. Compounds were isolated by repeated column chromatography on silica gel. Their structures were elucidated on the basis of spectral analysis and comparison with literature data. Six compounds were isolated and identified as codonolactone (1), voleneol (2), octacosanoic acid (3), beta-sitosterol (4), mangdesisterol (5), and daucosterol (6). Compounds 1, 2, and 5 were isolated from the plant for the first time.
Sigma-Aldrich
Octacosanoic acid, synthetic, ≥98%
Pricing and availability is not currently available.