Merck
  • Synthesis and characterization of branched polymers from lipase-catalyzed trimethylolpropane copolymerizations.
Synthesis and characterization of branched polymers from lipase-catalyzed trimethylolpropane copolymerizations.
Biomacromolecules (2007-05-05)
Ankur S Kulshrestha, Wei Gao, Hongyong Fu, Richard A Gross
ABSTRACT
Lipase-catalyzed terpolymerizations were performed with the monomers trimethylolpropane (B3), 1,8-octanediol (B2), and adipic acid (A2). Polymerizations were performed in bulk, at 70 degrees C, for 42 h, using immobilized lipase B from Candida antartica (Novozyme-435) as a catalyst. To determine the substitution pattern of trimethylolpropane (TMP) in copolymers, model compounds with variable degrees of acetylation were synthesized. Inverse-gated 13C NMR spectra were recorded to first determine the chemical shift positions for mono-, di-, and trisubstituted TMP units and, subsequently, to determine substitution of TMP units along chains. Variation of TMP in the monomer feed gave copolymers with degrees of branching (DB) from 20% to 67%. In one example, a hyperbranched copolyester with 53 mol % TMP adipate units was formed in 80% yield, with Mw 14 100 (relative to polystyrene standards), Mw/Mn 5.3, and DB 36%. Thermal and crystalline properties of the copolyesters were studied by thermogravimetric analysis and differential scanning calorimetry.
Sigma-Aldrich
Di(trimethylolpropane), 97%
Sigma-Aldrich
1,1,1-Tris(hydroxymethyl)propane, 97%
Sigma-Aldrich
1,1,1-Tris(hydroxymethyl)propane, dist., ≥98.0% (GC)