Merck
AP-1 activation mediates post-natal cardiomyocyte maturation.
Cardiovascular research (2022-06-01)
Hongjie Zhang, Lijuan Pei, Zhaohui Ouyang, Haocun Wang, Xin Chen, Kai Jiang, Shiqi Huang, Rui Jiang, Yaozu Xiang, Ke Wei
ABSTRACT
Post-natal maturation of mammalian cardiomyocytes proceeds rapidly after birth, with most of the myocytes exiting cell cycle, becoming binucleated, and adopting oxidative phosphorylation as the primary metabolic route. The triggers and transcriptional programmes regulating cardiomyocyte maturation have not been fully understood yet. We performed single-cell RNA-Seq in post-natal rat hearts in order to identify the important factors for this process. Single-cell RNA-Seq profiling was performed of post-natal Day 1 and Day 7 rat hearts, and we found that members of the activating protein 1 (AP-1) transcription factors showed a transient up-regulation in the maturing cardiomyocytes, suggesting their functional involvement in the process. Activating members of the AP-1 family by palmitate or adrenergic stimulation inhibited cardiomyocyte cytokinesis and promoted cardiomyocyte maturation. In contrast, knocking down AP-1 members Atf3 and Jun promoted cardiomyocyte cytokinesis, reduced polyploidy, and inhibited maturation. Mechanistically, RNA-Seq results and rescue experiments indicated that AP-1 members activate the expression of fatty acid metabolic genes to promote cardiomyocyte maturation. Finally, intraperitoneal injection of AP-1 inhibitor T-5224 in neonatal mice inhibits cardiomyocyte maturation in vivo. Our results are the first evidence implicating AP-1 transcription factors in post-natal cardiomyocyte maturation both in vitro and in vivo, which expand our understanding of the molecular mechanism of cardiomyocyte maturation, and may lead to novel therapies to treat congenital heart diseases.
Sigma-Aldrich
T-5224, ≥98% (HPLC)