Merck
  • Reticular groove of the domestic ruminants: histochemical and immunocytochemical study.
Reticular groove of the domestic ruminants: histochemical and immunocytochemical study.
Anatomia, histologia, embryologia (2012-04-18)
G Scala, L Maruccio
ABSTRACT
The reticular groove mucosa of adult cattle, buffalo and sheep was investigated by histochemical and immunocytochemical techniques. Intense NADPH-d staining was observed in the folds of the epithelium mucosa and at the bottom of the reticular groove in all domestic ruminants studied. The NADPH-d staining showed that the innervations of the tunica muscularis of the reticular groove lip were composed of nerve corpuscles, nerve fibres and nerve cells of the mucosa epithelium. SEM analysis showed an intense nitric oxide synthase (NOS) I immunoreactivity in deep and medium cellular layers. It is interesting to note that the same morphologies were observed in samples of the mucosa epithelium, and of the tunica muscularis processed by NADPH-d and in those processed by immunogold techniques. This study has demonstrated that nitric oxide (NO) is involved in the rumination activity and that it plays a double role in this activity in the reticular groove of all domestic ruminants studied: (1) NO plays a role similar to the one it has in the mucosa epithelium of all the other compartments of the ruminant forestomach, (2) The lip sections of the reticular groove has shown abundant innervations that may indirectly coordinate and control the forestomach motility through the direct activation of the nitrergic (nitroxidergic) nerve cells and nerve fibres.
Sigma-Aldrich
Anti-Nitric Oxide Synthase I Antibody, Chemicon®, from rabbit