Merck
  • Identification of Natural Product Sulfuretin Derivatives as Inhibitors for the Endoplasmic Reticulum Redox Protein ERO1α.
Identification of Natural Product Sulfuretin Derivatives as Inhibitors for the Endoplasmic Reticulum Redox Protein ERO1α.
ACS bio & med chem Au (2022-07-28)
Brennan D Johnson, Sridhar Kaulagari, Wei-Chih Chen, Karen Hayes, Werner J Geldenhuys, Lori A Hazlehurst
ABSTRACT
The flavin adenine dinucleotide containing Endoplasmic Reticulum Oxidoreductase-1 α (ERO1α) catalyzes the formation of de novo disulfide bond formation of secretory and transmembrane proteins and contributes towards proper protein folding. Recently, increased ERO1α expression has been shown to contribute to increased tumor growth and metastasis in multiple cancer types. In this report we sought to define novel chemical space for targeting ERO1α function. Using the previously reported ERO1α inhibitor compound, EN-460, as a benchmark pharmacological tool we were able to identify a sulfuretin derivative, T151742 which was approximately two-fold more potent using a recombinant enzyme assay system (IC50 = 8.27 ± 2.33 μM) compared to EN-460 (IC50= 16.46 ± 3.47 μM). Additionally, T151742 (IC50 = 16.04 μM) was slightly more sensitive than EN-460 (IC50= 19.35μM) using an MTT assay as an endpoint. Utilizing a cellular thermal shift assay (CETSA), we determined that the sulfuretin derivative T151742 demonstrated isozyme specificity for ERO1α as compared to ERO1β and showed no detectable binding to the FAD containing enzyme LSD-1. T151742 retained activity in PC-9 cells in a clonogenicity assay while EN-460 was devoid of activity. Furthermore, the activity of T151742 inhibition of clonogenicity was dependent on ERO1α expression as CRISPR edited PC-9 cells were resistant to treatment with T151742. In summary we identified a new scaffold that shows specificity for ERO1α compared to the closely related paralog ERO1β or the FAD containing enzyme LSD-1 that can be used as a tool compound for inhibition of ERO1α to allow for pharmacological validation of the role of ERO1α in cancer.
Sigma-Aldrich
ERO1 Inhibitor II, EN460
Sigma-Aldrich
LSD1 Inhibitor IV, RN-1, 2HCl, LSD1 Inhibitor IV, RN-1, HCl, is a cell-permeable potent, irreversible inhibitor of lysine specific demethylase 1 (LSD1; IC₅₀ = 70 nM in a HRP-coupled assay using H3K4Me2 peptide substrate).