Merck
  • An allosteric site in the T-cell receptor Cβ domain plays a critical signalling role.
An allosteric site in the T-cell receptor Cβ domain plays a critical signalling role.
Nature communications (2017-05-17)
Kannan Natarajan, Andrew C McShan, Jiansheng Jiang, Vlad K Kumirov, Rui Wang, Huaying Zhao, Peter Schuck, Mulualem E Tilahun, Lisa F Boyd, Jinfa Ying, Ad Bax, David H Margulies, Nikolaos G Sgourakis
ABSTRACT
The molecular mechanism through which the interaction of a clonotypic αβ T-cell receptor (TCR) with a peptide-loaded major histocompatibility complex (p/MHC) leads to T-cell activation is not yet fully understood. Here we exploit a high-affinity TCR (B4.2.3) to examine the structural changes that accompany binding to its p/MHC ligand (P18-I10/H2-Dd). In addition to conformational changes in complementarity-determining regions (CDRs) of the TCR seen in comparison of unliganded and bound X-ray structures, NMR characterization of the TCR β-chain dynamics reveals significant chemical shift effects in sites removed from the MHC-binding site. Remodelling of electrostatic interactions near the Cβ H3 helix at the membrane-proximal face of the TCR, a region implicated in interactions with the CD3 co-receptor, suggests a possible role for an allosteric mechanism in TCR signalling. The contribution of these TCR residues to signal transduction is supported by mutagenesis and T-cell functional assays.
Sigma-Aldrich
2-Keto-3-(methyl-d3)-butyric acid-1,2,3,4-13C4, 3-d sodium salt, 99 atom % 13C, 98 atom % D, ≥99% (CP)