Merck
  • Novel Crizotinib-GnRH Conjugates Revealed the Significance of Lysosomal Trapping in GnRH-Based Drug Delivery Systems.
Novel Crizotinib-GnRH Conjugates Revealed the Significance of Lysosomal Trapping in GnRH-Based Drug Delivery Systems.
International journal of molecular sciences (2019-11-14)
József Murányi, Attila Varga, Pál Gyulavári, Kinga Pénzes, Csilla E Németh, Miklós Csala, Lilla Pethő, Antal Csámpai, Gábor Halmos, István Peták, István Vályi-Nagy
ABSTRACT
Several promising anti-cancer drug-GnRH (gonadotropin-releasing hormone) conjugates have been developed in the last two decades, although none of them have been approved for clinical use yet. Crizotinib is an effective multi-target kinase inhibitor, approved against anaplastic lymphoma kinase (ALK)- or ROS proto-oncogene 1 (ROS-1)-positive non-small cell lung carcinoma (NSCLC); however, its application is accompanied by serious side effects. In order to deliver crizotinib selectively into the tumor cells, we synthesized novel crizotinib analogues and conjugated them to a [d-Lys6]-GnRH-I targeting peptide. Our most prominent crizotinib-GnRH conjugates, the amide-bond-containing [d-Lys6(crizotinib*)]-GnRH-I and the ester-bond-containing [d-Lys6(MJ55*)]-GnRH-I, were able to bind to GnRH-receptor (GnRHR) and exert a potent c-Met kinase inhibitory effect. The efficacy of compounds was tested on the MET-amplified and GnRHR-expressing EBC-1 NSCLC cells. In vitro pharmacological profiling led to the conclusion that that crizotinib-GnRH conjugates are transported directly into lysosomes, where the membrane permeability of crizotinib is diminished. As a consequence of GnRHR-mediated endocytosis, GnRH-conjugated crizotinib bypasses its molecular targets-the ATP-binding site of RTKs- and is sequestered in the lysosomes. These results explained the lower efficacy of crizotinib-GnRH conjugates in EBC-1 cells, and led to the conclusion that drug escape from the lysosomes is a major challenge in the development of clinically relevant anti-cancer drug-GnRH conjugates.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Sigma-Aldrich
Anti-Galectin 3 antibody produced in rabbit, affinity isolated antibody