Skip to Content
Merck
Zolpidem tartrate British Pharmacopoeia (BP) Reference Standard

Zolpidem tartrate

Zolpidem tartrate British Pharmacopoeia (BP) Reference Standard
Empirical Formula (Hill Notation):
C38H42N6O2 · C4H6O6
CAS Number:
Molecular Weight:
764.87

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Application
Package Size
API Family
Reference Material Type
Special Grade