
Sodium acetate trihydrate indifferent to potassium permanganate

Synonym(s):
Sodium acetate trihydrate, Acetic acid sodium salt trihydrate
Linear Formula:
CH3COONa · 3H2O
CAS Number:
Molecular Weight:
136.08
Products
Brand
Formula Weight
Showing 1-1 of 1
Compare
Product Number
Description
Pricing