
PCPDTBT

Synonym(s):
Poly[2,6-(4,4-bis-(2-ethylhexyl)-4H-cyclopenta [2,1-b;3,4-b′]dithiophene)-alt-4,7(2,1,3-benzothiadiazole)]
Linear Formula:
(C31H38N2S3)n
CAS Number:
Products
Brand
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing