
MGK 264

Synonym(s):
N-(2-Ethylhexyl)-5-norbornene-2,3-dicarboximide
Empirical Formula (Hill Notation):
C17H25NO2
CAS Number:
Molecular Weight:
275.39
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2