
Methiocarb sulfoxide

Synonym(s):
4-Methylsulfinyl-3,5-xylyl-N-methylcarbamate, Mercaptodimethur sulfoxide, Mesurol sulfoxide, Methiocarb breakdown product
Empirical Formula (Hill Notation):
C11H15NO3S
CAS Number:
Molecular Weight:
241.31
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2