Skip to Content
Merck
m-Cresol SAJ first grade, ≥97.0%

m-Cresol

m-Cresol SAJ first grade, ≥97.0%
Synonym(s):
1-Hydroxy-3-methylbenzene, 3-Hydroxytoluene, 3-Methylphenol, m-Cresol
Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing