
Felbinac impurity A

Synonym(s):
4-Acetylbiphenyl, 4-Biphenyl methyl ketone, 4-Phenylacetophenone
Linear Formula:
C6H5C6H4COCH3
CAS Number:
Molecular Weight:
196.24
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing