
Dipyridamole impurity standard

Synonym(s):
Dipyridamole
Empirical Formula (Hill Notation):
C24H40N8O4
CAS Number:
Molecular Weight:
504.63
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing