
4-Aminobenzoic acid

Synonym(s):
PABA, Vitamin Bx, Vitamin H1, 4-Aminobenzoic acid
Linear Formula:
H2NC6H4CO2H
CAS Number:
Molecular Weight:
137.14
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing