3-(3-Hydroxypropyl)-1H-indole
Synonym(s):
Homotryptophol, 3-Indolepropanol
Empirical Formula (Hill Notation):
C11H13NO
CAS Number:
Molecular Weight:
175.23
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing