Skip to Content
Merck
All Photos(1)

Key Documents

PHL89874

Angelicin

phyproof® Reference Substance

Synonym(s):

2-Oxo-(2H)-furo(2,3-h)-1-benzopyran, 2H-Furo[2,3-h]-1-benzopyran-2-one, Isopsoralen, NSC 404563

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H6O3
CAS Number:
Molecular Weight:
186.16
Beilstein:
153970
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥98.0% (HPLC)

form

solid

manufacturer/tradename

PhytoLab

storage temp.

2-8°C

SMILES string

O=C1Oc2c(C=C1)ccc3occc23

InChI

1S/C11H6O3/c12-10-4-2-7-1-3-9-8(5-6-13-9)11(7)14-10/h1-6H

InChI key

XDROKJSWHURZGO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Biochem/physiol Actions

Angular furocoumarin with diverse photobiological effects. Upon long-wavelength UV irradiation, forms monoadduct with double-stranded DNA and reacts with unsaturated fatty acids. Inhibits DNA and RNA synthesis and cell replication in Ehrlich ascites tumor cells.

Other Notes

This compound is commonly found in plants of the genus: angelica pimpinella

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service