Skip to Content
Merck
All Photos(1)

Key Documents

BP836

Venlafaxine impurity standard

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

Venlafaxine hydrochloride, (+/-)-1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol hydrochloride, Effexor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO2 · HCl
CAS Number:
Molecular Weight:
313.86
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

venlafaxine

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

mixture

storage temp.

2-8°C

SMILES string

Cl[H].COc1ccc(cc1)C(CN(C)C)C2(O)CCCCC2

InChI

1S/C17H27NO2.ClH/c1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14;/h7-10,16,19H,4-6,11-13H2,1-3H3;1H

InChI key

QYRYFNHXARDNFZ-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Venlafaxine impurity standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Also used in monographs such as:
  • Venlafaxine Tablets
  • Venlafaxine Prolonged-release Tablets
  • Venlafaxine Prolonged-release Capsules
  • Biochem/physiol Actions

    Venlafaxine is an antidepressant. The mechanism of the antidepresant action of venlafaxine in humans is associated with its potentiation of neurotransmitter activity in the CNS. Venlafaxine is a potent inhibitor of neuronal serotonin and norepinephrine reuptake and weak inhibitor of dopamine reuptake. Venlafaxine has no significant activity for muscarinic, histaminergic, or α-1 adrenergic receptors in vitro. Venlafaxine does not possess MAO inhibitor activity.

    Packaging

    Unit quantity: 25 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

    Other Notes

    Sales restrictions may apply.

    Pictograms

    Environment

    Hazard Statements

    Precautionary Statements

    Hazard Classifications

    Aquatic Chronic 2

    Storage Class Code

    11 - Combustible Solids

    WGK

    WGK 3

    Flash Point(F)

    Not applicable

    Flash Point(C)

    Not applicable


    Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service