Key Documents
TMO362150-1130
Nalgene® white caps
polypropylene, diam. × thread 16 mm × 13-415 mm, case of 2000 ea
Synonym(s):
leakproof closure, threaded cap
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
polypropylene
white
packaging
case of 2000 ea
manufacturer/tradename
Nalgene 362150-1130
diam. × thread
16 mm × 13-415 mm
General description
Eliminate spills with Thermo Scientific Nalgene Colored Polypropylene Closures. These one-piece linerless closures fit Nalgene narrow-mouth 4mL bottles. They feature an integrally molded seal ring and a well-defined closure knurl that makes gripping and capping easy.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries