Improved HPLC Selectivity of Discovery® BIO Wide Pore RP Phases
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery BIO Wide Pore C18, 15 cm x 4.6 mm I.D., 5 μm particles vs. competitive wide pore C18 column of same dimensions (568222-U)
mobile phase
[A] 0.1% TFA in 80:20 water:acetonitrile; [B] 0.1% TFA in 66:34 water:acetonitrile
Gradient of 0 to 100% B in 14 min. after 1 min. delay
gradient
0 to 100% B in 14 min. after 1 min. delay
flow rate
1 mL/min
column temp.
30 °C
detector
UV, 220 nm
injection
10 μL
sample
peptide mixture (Sigma P2693), ~0.25 μg each peptide in 0.1% TFA
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany