Merck
  • Nanoparticles Embedded in Amphiphilic Membranes for Carbon Dioxide Separation and Dehumidification.
Nanoparticles Embedded in Amphiphilic Membranes for Carbon Dioxide Separation and Dehumidification.
ChemSusChem (2017-08-24)
Wai Fen Yong, Yan Xun Ho, Tai-Shung Chung
ABSTRACT
Polymers containing ethylene oxide (EO) groups have gained significant interest as the EO groups have favorable interactions with polar molecules such as H
Sigma-Aldrich
4,5-Dichloroimidazole, ≥98%
Pricing and availability is not currently available.