Merck
  • Soluble Platinum Nanoparticles Ligated by Long-Chain N-Heterocyclic Carbenes as Catalysts.
Soluble Platinum Nanoparticles Ligated by Long-Chain N-Heterocyclic Carbenes as Catalysts.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-06-15)
Luis M Martínez-Prieto, Lena Rakers, Angela M López-Vinasco, Israel Cano, Yannick Coppel, Karine Philippot, Frank Glorius, Bruno Chaudret, Piet W N M van Leeuwen
ABSTRACT
Soluble platinum nanoparticles (Pt NPs) ligated by two different long-chain N-heterocyclic carbenes (LC-IPr and LC-IMe) were synthesized and fully characterized by TEM, high-resolution TEM, wide-angle X-ray scattering (WAXS), X-ray photoelectron spectroscopy (XPS), and solution NMR. The surface chemistry of these NPs (Pt@LC-IPr and Pt@LC-IMe) was investigated by FT-IR and solid state NMR using CO as a probe molecule. A clear influence of the bulkiness of the N-substituents on the size, surface state, and catalytic activity of these Pt NPs was observed. While Pt@LC-IMe showed no activity in the hydroboration of phenylacetylene, Pt@LC-IPr revealed good selectivity for the trans-isomer, which may be supported by a homogeneous species. This is the first example of hydroboration of acetylenes catalyzed by non-supported Pt NPs.
Sigma-Aldrich
1-Ethynyl-4-fluorobenzene, 99%
Sigma-Aldrich
4-Ethynylanisole, 97%