Merck
Coumarin Based Fluorescent Probe for Colorimetric Detection of Fe
Journal of fluorescence (2017-03-17)
Nayan Roy, Abhijit Dutta, Paritosh Mondal, Pradip C Paul, T Sanjoy Singh
ABSTRACT
A new coumarin based Schiff-base chemosensor-(E)-7-(((8-hydroxyquinolin-2-yl)methylene) amino)-4-methyl-2H-chromen-2-one (H
Sigma-Aldrich
8-Hydroxy-2-quinolinecarboxaldehyde, ≥98.0% (GC)