Merck
  • A Genomic DNA Reporter Screen Identifies Squalene Synthase Inhibitors That Act Cooperatively with Statins to Upregulate the Low-Density Lipoprotein Receptor.
A Genomic DNA Reporter Screen Identifies Squalene Synthase Inhibitors That Act Cooperatively with Statins to Upregulate the Low-Density Lipoprotein Receptor.
The Journal of pharmacology and experimental therapeutics (2017-04-01)
Alastair G Kerr, Lawrence C S Tam, Ashley B Hale, Milena Cioroch, Gillian Douglas, Sarina Agkatsev, Olivia Hibbitt, Joseph Mason, James Holt-Martyn, Carole J R Bataille, Graham M Wynne, Keith M Channon, Angela J Russell, Richard Wade-Martins
ABSTRACT
Hypercholesterolemia remains one of the leading risk factors for the development of cardiovascular disease. Many large double-blind studies have demonstrated that lowering low-density lipoprotein (LDL) cholesterol using a statin can reduce the risk of having a cardiovascular event by approximately 30%. However, despite the success of statins, some patient populations are unable to lower their LDL cholesterol to meet the targeted lipid levels, due to compliance or potency issues. This is especially true for patients with heterozygous familial hypercholesterolemia who may require additional upregulation of the low-density lipoprotein receptor (LDLR) to reduce LDL cholesterol levels below those achievable with maximal dosing of statins. Here we identify a series of small molecules from a genomic DNA reporter screen that upregulate the LDLR in mouse and human liver cell lines at nanomolar potencies (EC
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
OX03050, ≥98% (HPLC)