Merck
  • Application of the aza-Achmatowicz oxidative rearrangement for the stereoselective synthesis of the Cassia and Prosopis alkaloid family.
Application of the aza-Achmatowicz oxidative rearrangement for the stereoselective synthesis of the Cassia and Prosopis alkaloid family.
The Journal of organic chemistry (2006-10-27)
Carolyn A Leverett, Michael P Cassidy, Albert Padwa
ABSTRACT
cis-2-Methyl-6-substituted piperidin-3-ol alkaloids of the Cassia and Prosopis species are readily prepared by a combination of an aza-Achmatowicz oxidative rearrangement and dihydropyridone reduction followed by a stereoselective allylsilane addition to a N-sulfonyliminium ion. The stereochemical outcome of the reduction reaction can be attributed to steric hindrance between the pseudoaxially oriented 2,6-substituents and the equatorially approaching hydride reagent which explains the exclusive formation of the cis-alcohol by axial approach of the hydride. The unsaturation present in the (E)-methyl-pent-3-enoate side chain was removed by catalytic reduction, and the remaining ester group was converted to the corresponding Weinreb's amide. This key intermediate was utilized for the synthesis of azimic acid, deoxocassine, cassine, and spicigerine. The facile preparation of (S)-N-tosylamidofuran 16 and its conversion to the chiral Achmatowicz oxidation product 18 provide a formal chiral synthesis of these alkaloids.
Sigma-Aldrich
8-Bromo-1-octene, 97%
Pricing and availability is not currently available.