Merck
  • Effect of Fibulin-5 on cell proliferation and invasion in human gastric cancer patients.
Effect of Fibulin-5 on cell proliferation and invasion in human gastric cancer patients.
Asian Pacific journal of tropical medicine (2014-08-19)
Xiao-Yu Shi, Liang Wang, Chun-Hui Cao, Zhi-Yu Li, Jian Chen, Chen Li
ABSTRACT
To explore the effect of Fibulin-5 expression on cell proliferation and invasion in human gastric cancer patients. Fibulin-5 expression was detected in 56 samples of surgically resected gastric cancer and paired noncancerous tissues using qRT-PCR and immunoblotting. Fibulin-5 was knocked down by Fibulin-5 shRNA in MGC-803 cells, then BrdU cell proliferation and transwell invasion assays were used to determine cell proliferation and invasion. The level of Fibulin-5 mRNA in gastric cancer tissues was significantly higher as compared with that in normal tumor-adjacent tissues (P<0.05). Otherwise, the level of Fibulin-5 protein in cancer and noncancerous tissues was consistent with mRNA expression (P<0.05). Fibulin-5 protein expression in tumor tissues with poorly differentiated, lymph node metastasis and advanced TNM tumor stage was significantly higher (P<0.05, respectively). Fibulin-5 was obviously knocked down by Fibulin-5 shRNA (P<0.05), and Fibulin-5 knockdown significantly inhibited cell proliferation and invasion in MGC-803 cells (P<0.05, respectively). The high-expression of Fibulin-5 is associated with the malignant clinicopathologic parameters in gastric cancer and Fibulin-5 knockdown inhibits cell proliferation and invasion in MGC-803 cells, suggesting Fibulin-5 may act as a key factor in the progression of gastric cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human NTS
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fbln5
Sigma-Aldrich
MISSION® esiRNA, targeting human FBLN5
Sigma-Aldrich
MISSION® esiRNA, targeting human NT5E