- Mead acid inhibits the growth of KPL-1 human breast cancer cells in vitro and in vivo.
Mead acid inhibits the growth of KPL-1 human breast cancer cells in vitro and in vivo.
The effects of mead acid (MA; 5,8,11-eicosatrienoic acid) on the suppression of breast cancer cell growth and metastasis were examined inĀ vitro and inĀ vivo by using the KPL-1 human breast cancer cell line. MA suppressed KPL-1 cell growth in culture with an IC50 value of 214.2Ā ĀµM (65.7Ā Āµg/ml) for 72Ā h, and MA significantly suppressed transplanted KPL-1 tumor growth (tumor volume and tumor weight: 872Ā±103Ā mm3 and 1,000Ā±116Ā mg vs. 376Ā±66 mm3 and 517Ā±84Ā mg) and regional (axillary) lymph node metastasis (67%, 10/15 vs. 10%, 1/10) in female athymic mice fed an MA-rich diet for 8Ā weeks. Tumor suppression was due to the suppression of cell proliferation. In ELISA, although vascular endothelial growth factor (VEGF) levels were unchanged, VEGF receptor (VEGFR)1 and VEGFR2 levels were significantly decreased after treatment with a 214.2-ĀµM dose of MA for 72Ā h; E-cadherin levels were unchanged. As VEGF, VEGFR1 and VEGFR2 expression was co-localized in KPL-1 cells, the mechanism leading to cell growth suppression was VEGF signaling directly to KPL-1 cells by an autocrine process. In contrast, MA did not influence angiogenesis. The mechanisms of action were through VEGF signaling directly to cancer cells.