Merck
Engineered biosynthesis of medium-chain esters in Escherichia coli.
Metabolic engineering (2014-12-03)
Yi-Shu Tai, Mingyong Xiong, Kechun Zhang
ABSTRACT
Medium-chain esters such as isobutyl acetate (IBAc) and isoamyl acetate (IAAc) are high-volume solvents, flavors and fragrances. In this work, we engineered synthetic metabolic pathways in Escherichia coli for the total biosynthesis of IBAc and IAAc directly from glucose. Our pathways harnessed the power of natural amino acid biosynthesis. In particular, the native valine and leucine pathways in E. coli were utilized to supply the precursors. Then alcohol acyltransferases from various organisms were investigated on their capability to catalyze esterification reactions. It was discovered that ATF1 from Saccharomyces cerevisiae was the best enzyme for the formation of both IBAc and IAAc in E. coli. In vitro biochemical characterization of ATF1 confirmed the fermentation results and provided rational guidance for future enzyme engineering. We also performed strain improvement by removing byproduct pathways (Δldh, ΔpoxB, Δpta) and increased the production of both target chemicals. Then the best IBAc producing strain was used for scale-up fermentation in a 1.3-L benchtop bioreactor. 36g/L of IBAc was produced after 72h fermentation. This work demonstrates the feasibility of total biosynthesis of medium-chain esters as renewable chemicals.
Sigma-Aldrich
Isobutyl acetate, FCC, FG
Sign Into View Organizational & Contract Pricing
SKUVendor SKUPack SizeAvailabilityPrice
EN300-1627788-100MG100 mg
Ships in 7 days - from Aldrich Partner.
$327.60
EN300-1627788-250MG250 mg
Ships in 7 days - from Aldrich Partner.
$468.00
EN300-1627788-500MG500 mg
Ships in 7 days - from Aldrich Partner.
$738.40
EN300-1627788-1G1 g
Ships in 7 days - from Aldrich Partner.
$946.40
EN300-1627788-2.5G2.5 g
Ships in 7 days - from Aldrich Partner.
$1,856.40
EN300-1627788-5G5 g
Ships in 7 days - from Aldrich Partner.
$2,743.00
EN300-1627788-10G10 g
Ships in 7 days - from Aldrich Partner.
$4,069.00
To order products, please contact your local dealer.