Merck
  • Chemopreventive properties of chlorophylls towards aflatoxin B1: a review of the antimutagenicity and anticarcinogenicity data in rainbow trout.
Chemopreventive properties of chlorophylls towards aflatoxin B1: a review of the antimutagenicity and anticarcinogenicity data in rainbow trout.
Mutation research (1998-07-22)
R Dashwood, T Negishi, H Hayatsu, V Breinholt, J Hendricks, G Bailey
ABSTRACT
The anticarcinogenic activity of chlorophyllin (CHL), a water-soluble derivative of chlorophyll, was first reported in rainbow trout. This review describes certain experiments which set the stage for long-term tumor bioassays, in trout and other species, using CHL and various food-borne carcinogens. Initial work with trout and rat liver enzymes in the Salmonella assay showed that CHL was a potent antimutagen towards heterocyclic amines, polycyclic aromatic hydrocarbons, aflatoxins and other classes of mutagen. Antimutagenic activity was further demonstrated using the corresponding direct-acting mutagens in the absence of an exogenous metabolizing system. Mutagen-inhibitor interaction (molecular complex formation) was identified in spectrophotometry studies, suggesting that CHL acts as an 'interceptor molecule'. In vivo, CHL reduced hepatic AFB1-DNA adducts and hepatocarcinogenesis when the inhibitor and carcinogen were co-administered in the diet. Finally, co-injection of inhibitor and AFB1 into trout embryos established that CHL was more effective than chlorophyll a in reducing AFB1-DNA adducts 2 weeks after injection, and liver tumors after 1 year.
Sigma-Aldrich
Aflatoxin B1 from Aspergillus flavus, from Aspergillus flavus
Supelco
Aflatoxin B1 solution, 2 μg/mL in acetonitrile, analytical standard
Aflatoxin B1 solution, 3.79 μg/g in acetonitrile, ERM®, certified reference material
Supelco
Aflatoxin B1 solution, 3 μg/mL in benzene:acetonitrile (98:2), analytical standard