Merck
  • Baicalin as a potentially promising drug for the management of sulfur mustard induced cutaneous complications: a review of molecular mechanisms.
Baicalin as a potentially promising drug for the management of sulfur mustard induced cutaneous complications: a review of molecular mechanisms.
Cutaneous and ocular toxicology (2011-11-24)
Amirhossein Sahebkar
ABSTRACT
Sulfur mustard (SM) is a bifunctional alkylating agent with strong blistering, irritant, mutagenic and cytotoxic properties. SM has been widely deployed as a chemical warfare agent for over a century, leading to extensive casualties. Skin is among the first and most heavily damaged organs upon SM exposure. Unfortunately, a considerable fraction of SM-intoxicated patients are still suffering from chronic cutaneous complications. While these complications adversely affect patients' quality of life, there is as yet no ideal treatment for them and therapeutic options are limited and mainly symptomatic. During recent decades, remarkable progress has been made in understanding molecular mechanisms underlying SM-induced dermatotoxicity and several intra- and extracellular targets have been identified. This review argues that baicalin, a bioactive flavonoid from the roots of Scutellaria spp., could counteract different molecular and biochemical abnormalities that mediate SM dermatotoxicity and could therefore be regarded as a promising therapeutic option for the management of SM-induced cutaneous lesions.
Product Description
Baicalin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Baicalin, 95%