Merck
  • Randomized, placebo-controlled, phase III trial of sunitinib plus prednisone versus prednisone alone in progressive, metastatic, castration-resistant prostate cancer.
Randomized, placebo-controlled, phase III trial of sunitinib plus prednisone versus prednisone alone in progressive, metastatic, castration-resistant prostate cancer.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2013-12-11)
M Dror Michaelson, Stephane Oudard, Yen-Chuan Ou, Lisa Sengeløv, Fred Saad, Nadine Houede, Peter Ostler, Arnulf Stenzl, Gedske Daugaard, Robert Jones, Fredrik Laestadius, Anders Ullèn, Amit Bahl, Daniel Castellano, Juergen Gschwend, Tristan Maurina, Edna Chow Maneval, Shaw-Ling Wang, Maria Jose Lechuga, Jolanda Paolini, Isan Chen
ABSTRACT
We evaluated angiogenesis-targeted sunitinib therapy in a randomized, double-blind trial of metastatic castration-resistant prostate cancer (mCRPC). Men with progressive mCRPC after docetaxel-based chemotherapy were randomly assigned 2:1 to receive sunitinib 37.5 mg/d continuously or placebo. Patients also received oral prednisone 5 mg twice daily. The primary end point was overall survival (OS); secondary end points included progression-free survival (PFS). Two interim analyses were planned. Overall, 873 patients were randomly assigned to receive sunitinib (n = 584) or placebo (n = 289). The independent data monitoring committee stopped the study for futility after the second interim analysis. After a median overall follow-up of 8.7 months, median OS was 13.1 months and 11.8 months for sunitinib and placebo, respectively (hazard ratio [HR], 0.914; 95% CI, 0.762 to 1.097; stratified log-rank test, P = .168). PFS was significantly improved in the sunitinib arm (median 5.6 v 4.1 months; HR, 0.725; 95% CI, 0.591 to 0.890; stratified log-rank test, P < .001). Toxicity and rates of discontinuations because of adverse events (AEs; 27% v 7%) were greater with sunitinib than placebo. The most common treatment-related grade 3/4 AEs were fatigue (9% v 1%), asthenia (8% v 2%), and hand-foot syndrome (7% v 0%). Frequent treatment-emergent grade 3/4 hematologic abnormalities were lymphopenia (20% v 11%), anemia (9% v 8%), and neutropenia (6% v < 1%). The addition of sunitinib to prednisone did not improve OS compared with placebo in docetaxel-refractory mCRPC. The role of antiangiogenic therapy in mCRPC remains investigational.
Product Description
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Sigma-Aldrich
Prednisone, ≥98%