Merck
  • Synthesis and antileishmanial activity of (1,3-benzothiazol-2-yl) amino-9-(10H)-acridinone derivatives.
Synthesis and antileishmanial activity of (1,3-benzothiazol-2-yl) amino-9-(10H)-acridinone derivatives.
European journal of medicinal chemistry (2004-07-28)
Florence Delmas, Antonio Avellaneda, Carole Di Giorgio, Maxime Robin, Erik De Clercq, Pierre Timon-David, Jean-Pierre Galy
ABSTRACT
(1,3-Benzothiazol-2-yl) amino-9-(10H)-acridinone derivatives were synthesized via a procedure based on the Ullman reaction and were assessed for their in vitro antileishmanial and anti-HIV activities. Two derivatives, 4-(6-nitro-benzothiazol-2-ylamino)-10H-acridin-9-one and 1-(6-amino-benzothiazol-2-ylamino)-10H-acridin-9-one, revealed a selective antileishmanial activity, mainly due to amastigote-specific toxicity. Results suggested that:the addition of a benzothiazole group on a parent amino-9-(10H)-acridinone ring could enhance antileishmanial abilities, the presence of a 6-amino-benzothiazole group on position 2 amino chain or a 6-nitro-benzothiazole group on position 4 amino chain was essential for specific anti-amastigote properties.
Sigma-Aldrich
2-Chlorobenzothiazole, 99%
Pricing and availability is not currently available.