Merck
  • Crystal structure of cyclophilin A complexed with substrate Ala-Pro suggests a solvent-assisted mechanism of cis-trans isomerization.
Crystal structure of cyclophilin A complexed with substrate Ala-Pro suggests a solvent-assisted mechanism of cis-trans isomerization.
Proceedings of the National Academy of Sciences of the United States of America (1993-04-15)
H Ke, D Mayrose, W Cao
ABSTRACT
Cyclophilin is a binding protein for the immunosuppressive drug cyclosporin A and is also an enzyme with peptidyl-prolyl cis-trans isomerase activity. The crystal structure of cyclophilin A complexed with the substrate Ala-Pro has been determined and refined to an R factor of 0.196 at 1.64-A resolution. The structure shows that only the cis form of Ala-Pro binds cyclophilin A despite the fact that Ala-Pro has an equilibrium majority of the trans form in solution. Simulation of the cis-trans isomerization in an ESV10 graphics system suggests a solvent-assisted mechanism in which first the peptidyl-prolyl bond is desolvated at the ground state by binding to the hydrophobic pocket of the active site, and later the intermediate state is stabilized by a hydrogen bond between the carbonyl oxygen of the amide bond and a bound water molecule.
Sigma-Aldrich
Ala-Pro hydrate
Pricing and availability is not currently available. Contact Technical Service