Merck
  • Computer-automated prediction of the mutagenicity of benzidine, 4,4"-diaminoterphenyl, 4-dimethylaminoazobenzene and 4-cyanodimethylaniline: comparison with the results of the Second UKEMS Collaborative Study.
Computer-automated prediction of the mutagenicity of benzidine, 4,4"-diaminoterphenyl, 4-dimethylaminoazobenzene and 4-cyanodimethylaniline: comparison with the results of the Second UKEMS Collaborative Study.
Mutagenesis (1986-07-01)
H S Rosenkranz, M R Frierson, G Klopman
ABSTRACT
There was agreement between the experimental results, obtained in the course of the Second UKEMS Collaborative Study, for the mutagenicity in Salmonella typhimurium of benzidine, 4,4"-diaminoterphenyl, 4-dimethylaminoazobenzene and 4-cyanodimethylaniline and the mutagenicity predicted by CASE (Computer Automated Structure Evaluation), a recently developed artificial intelligence system.
Sigma-Aldrich
4-(Dimethylamino)benzonitrile, 98%